Skip to Content
MilliporeSigma
Acitretin British Pharmacopoeia (BP) Reference Standard

Acitretin

Acitretin British Pharmacopoeia (BP) Reference Standard
Synonym(s):
9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid
Empirical Formula (Hill Notation):
C21H26O3
CAS Number:
Molecular Weight:
326.43
EC Number:

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Special Grade