Skip to Content
MilliporeSigma
Aceclofenac pharmaceutical secondary standard, certified reference material

Aceclofenac

Aceclofenac pharmaceutical secondary standard, certified reference material
Synonym(s):
2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid carboxymethyl ester;, Aceclofenac
Empirical Formula (Hill Notation):
C16H13Cl2NO4
CAS Number:
Molecular Weight:
354.18

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Agency
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2