Skip to Content
MilliporeSigma
α-(4-Pyridyl N-oxide)-N-tert-butylnitrone ~95%

α-(4-Pyridyl N-oxide)-N-tert-butylnitrone

α-(4-Pyridyl N-oxide)-N-tert-butylnitrone ~95%
Synonym(s):
N-(4-Pyridylmethylene)-tert-butylamine N,N′-dioxide, N-tert-Butyl-α-(4-pyridyl)nitrone N′-oxide, POBN
Empirical Formula (Hill Notation):
C10H14N2O2
CAS Number:
Molecular Weight:
194.23
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing