Skip to Content
MilliporeSigma
8-Bromooctanoic acid 97%

8-Bromooctanoic acid

8-Bromooctanoic acid 97%
Synonym(s):
8-Bromocaprylic acid
Linear Formula:
Br(CH2)7CO2H
CAS Number:
Molecular Weight:
223.11

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing