Skip to Content
MilliporeSigma
7-Azido-4-Methylcoumarin 97%

7-Azido-4-Methylcoumarin

7-Azido-4-Methylcoumarin 97%
Empirical Formula (Hill Notation):
C10H7N3O2
CAS Number:
Molecular Weight:
201.18

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing