Skip to Content
MilliporeSigma
6-Bromo-3,4-dihydro-2(1H)-naphthalenone 97%

6-Bromo-3,4-dihydro-2(1H)-naphthalenone

6-Bromo-3,4-dihydro-2(1H)-naphthalenone 97%
Synonym(s):
6-Bromo-2-tetralone
Empirical Formula (Hill Notation):
C10H9BrO
CAS Number:
Molecular Weight:
225.08

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing