Skip to Content
MilliporeSigma
6β-Hydroxytestosterone ≥97% (HPLC)

6β-Hydroxytestosterone

6β-Hydroxytestosterone ≥97% (HPLC)
Synonym(s):
4-Androstene-6β,17β-diol-3-one, 6β, 17β,-Dihydroxyandrost-4-en-3-one
Empirical Formula (Hill Notation):
C19H28O3
CAS Number:
Molecular Weight:
304.42

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing