Skip to Content
MilliporeSigma
5′-O-(4,4′-Dimethoxytrityl)thymidine 98%

5′-O-(4,4′-Dimethoxytrityl)thymidine

5′-O-(4,4′-Dimethoxytrityl)thymidine 98%
Synonym(s):
DMT-T
Empirical Formula (Hill Notation):
C31H32N2O7
CAS Number:
Molecular Weight:
544.59
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing