Skip to Content
MilliporeSigma
5-Methoxy-1-tetralone 97%

5-Methoxy-1-tetralone

5-Methoxy-1-tetralone 97%
Empirical Formula (Hill Notation):
C11H12O2
CAS Number:
Molecular Weight:
176.21
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing