Skip to Content
MilliporeSigma
5-Hydroxyindole-2-carboxylic acid ≥96.5%

5-Hydroxyindole-2-carboxylic acid

5-Hydroxyindole-2-carboxylic acid ≥96.5%
Synonym(s):
NSC 117338
Empirical Formula (Hill Notation):
C9H7NO3
CAS Number:
Molecular Weight:
177.16
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing