Skip to Content
MilliporeSigma
5α-Androst-16-en-3-one

5α-Androst-16-en-3-one

5α-Androst-16-en-3-one
Synonym(s):
16-(5α)Androsten-3-one, 3-Keto-5α,16-androstene, Androstenone
Empirical Formula (Hill Notation):
C19H28O
CAS Number:
Molecular Weight:
272.43
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing