Skip to Content
MilliporeSigma
5,8,11,14-Eicosatetraynoic acid ≥97%

5,8,11,14-Eicosatetraynoic acid

5,8,11,14-Eicosatetraynoic acid ≥97%
Synonym(s):
ETYA, Octadehydroarachidonic acid
Empirical Formula (Hill Notation):
C20H24O2
CAS Number:
Molecular Weight:
296.40

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing