Skip to Content
MilliporeSigma
5,6-trans-vitamin D3 ≥90% (HPLC)

5,6-trans-vitamin D3

5,6-trans-vitamin D3 ≥90% (HPLC)
Synonym(s):
5,6-trans-vitamin D3, (3b,5E,7E)-9,10-Secocholesta-5,7,10(19)-trien-3-ol, calciol, cholecalciferol, colecalciferol
Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing