Skip to Content
MilliporeSigma
5,6,7,8-Tetrahydroquinoxaline ≥97%, FG

5,6,7,8-Tetrahydroquinoxaline

5,6,7,8-Tetrahydroquinoxaline ≥97%, FG
Synonym(s):
Cyclohexapyrazine, Tetrahydroquinoxaline
Empirical Formula (Hill Notation):
C8H10N2
CAS Number:
Molecular Weight:
134.18
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing