Skip to Content
MilliporeSigma
4-(Trifluoromethyl)cyclohexanone 97%

4-(Trifluoromethyl)cyclohexanone

4-(Trifluoromethyl)cyclohexanone 97%
Empirical Formula (Hill Notation):
C7H9F3O
CAS Number:
Molecular Weight:
166.14

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing