Skip to Content
MilliporeSigma
4-tert-Butylbenzyl bromide 97%

4-tert-Butylbenzyl bromide

4-tert-Butylbenzyl bromide 97%
Synonym(s):
α-Bromo-4-(tert-butyl)toluene, 1-Bromomethyl-4-tert-butylbenzene
Linear Formula:
(CH3)3CC6H4CH2Br
CAS Number:
Molecular Weight:
227.14
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing