Skip to Content
MilliporeSigma
4-Methylumbelliferyl β-D-mannopyranoside ≥98% (TLC)

4-Methylumbelliferyl β-D-mannopyranoside

4-Methylumbelliferyl β-D-mannopyranoside ≥98% (TLC)
Synonym(s):
4-Methylumbelliferyl β-D-mannoside
Empirical Formula (Hill Notation):
C16H18O8
CAS Number:
Molecular Weight:
338.31

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing