Skip to Content
MilliporeSigma
4-(Methylamino)benzoic acid 97%

4-(Methylamino)benzoic acid

4-(Methylamino)benzoic acid 97%
Linear Formula:
CH3NHC6H4CO2H
CAS Number:
Molecular Weight:
151.16
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing