
4′-Methylacetophenone

Synonym(s):
Methyl p-tolyl ketone, 4′-Methylacetophenone, (1-(4-Methylphenyl)-1-ethanone (4′-Methylacetophenone))
Linear Formula:
CH3C6H4COCH3
CAS Number:
Molecular Weight:
134.18
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing