Skip to Content
MilliporeSigma
4-(Hydroxymethyl)benzonitrile 95%

4-(Hydroxymethyl)benzonitrile

4-(Hydroxymethyl)benzonitrile 95%
Synonym(s):
4-Cyanobenzyl alcohol
Linear Formula:
HOCH2C6H4CN
CAS Number:
Molecular Weight:
133.15

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing