Skip to Content
MilliporeSigma
4-Hydroxybenzaldehyde analytical standard

4-Hydroxybenzaldehyde

4-Hydroxybenzaldehyde analytical standard
Synonym(s):
4-Hydroxybenzaldehyde
Linear Formula:
HOC6H4CHO
CAS Number:
Molecular Weight:
122.12
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5