Skip to Content
MilliporeSigma
4-(Ethylaminomethyl)pyridine 97%

4-(Ethylaminomethyl)pyridine

4-(Ethylaminomethyl)pyridine 97%
Synonym(s):
N-(Pyridin-4-ylmethyl)ethanamine, N-Ethyl-4-pyridinemethanamine, NSC 85903
Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing