Skip to Content
MilliporeSigma
4-Bromophenylhydrazine hydrochloride 99%

4-Bromophenylhydrazine hydrochloride

4-Bromophenylhydrazine hydrochloride 99%
Linear Formula:
BrC6H4NHNH2·HCl
CAS Number:
Molecular Weight:
223.50

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing