Skip to Content
MilliporeSigma
4-Amino-1-benzylpiperidine 98%

4-Amino-1-benzylpiperidine

4-Amino-1-benzylpiperidine 98%
Synonym(s):
1-(Phenylmethyl)-4-piperidinamine
Empirical Formula (Hill Notation):
C12H18N2
CAS Number:
Molecular Weight:
190.28
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing