Skip to Content
AS
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Testing
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4,5-Dihydro-3(2
H
)-thiophenone
Synonym(s):
3-Thiophanone, Tetrahydrothiophen-3-one
Empirical Formula (Hill Notation):
C
4
H
6
OS
CAS Number:
1003-04-9
Molecular Weight:
102.15
EC Number:
213-698-9
Products
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
W326607
4,5-Dihydro-3(2
H
)-thiophenone,
≥98%, FG
Expand
View Pricing
264784
4,5-Dihydro-3(2
H
)-thiophenone,
98%
Expand
View Pricing