Skip to Content
MilliporeSigma
4-(4-Diethylaminostyryl)-1-methylpyridinium iodide ≥97%

4-(4-Diethylaminostyryl)-1-methylpyridinium iodide

4-(4-Diethylaminostyryl)-1-methylpyridinium iodide ≥97%
Synonym(s):
4-Di-2-ASP
Empirical Formula (Hill Notation):
C18H23IN2
CAS Number:
Molecular Weight:
394.29

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing