Skip to Content
MilliporeSigma
4,4′-Bis(diethylamino)benzophenone ≥99%

4,4′-Bis(diethylamino)benzophenone

4,4′-Bis(diethylamino)benzophenone ≥99%
Synonym(s):
4,4′-Bis(diethylamino)-benzophenone
Linear Formula:
4-[C2H5)2N]C6H4COC6H4-4-[N(C2H5)2]
CAS Number:
Molecular Weight:
324.46

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing