Skip to Content
MilliporeSigma
4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid) Dye content 75 %

4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid)

4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid) Dye content 75 %
Synonym(s):
meso-Tetraphenylporphine-4,4′,4″,4″′-tetracarboxylic acid
Empirical Formula (Hill Notation):
C48H30N4O8
CAS Number:
Molecular Weight:
790.77

Products

Product Category
Formula Weight
Physical Form
Chemical Composition
Markush Class
Markush Group
Showing 1-1 of 1