Skip to Content
MilliporeSigma
3-[Tris(trimethylsiloxy)silyl]propyl methacrylate contains MEHQ + HQ as stabilizer, 98%

3-[Tris(trimethylsiloxy)silyl]propyl methacrylate

3-[Tris(trimethylsiloxy)silyl]propyl methacrylate contains MEHQ + HQ as stabilizer, 98%
Synonym(s):
(3-Methacryloyloxypropyl)tris(trimethylsiloxy)silane
Linear Formula:
H2C=C(CH3)CO2(CH2)3Si[OSi(CH3)3]3
CAS Number:
Molecular Weight:
422.81
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1