Skip to Content
MilliporeSigma
3-((tert-Butyldimethylsilyl)oxy)-propanol 97%

3-((tert-Butyldimethylsilyl)oxy)-propanol

3-((tert-Butyldimethylsilyl)oxy)-propanol 97%
Linear Formula:
(CH3)3CSi(CH3)2OCH2CH2CH2OH
CAS Number:
Molecular Weight:
190.36

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing