Skip to Content
MilliporeSigma
3-O-Methyl-D-glucopyranose ≥98% (GC)

3-O-Methyl-D-glucopyranose

3-O-Methyl-D-glucopyranose ≥98% (GC)
Synonym(s):
3-O-Methyl-α-D-glucopyranose, 3-O-Methylglucose
Empirical Formula (Hill Notation):
C7H14O6
CAS Number:
Molecular Weight:
194.18
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing