Skip to Content
MilliporeSigma
3-Nitrophenylboronic acid ≥97%

3-Nitrophenylboronic acid

3-Nitrophenylboronic acid ≥97%
Synonym(s):
3-Nitrobenzeneboronic acid, m-Nitrobenzeneboronic acid, m-Nitrophenylboronic acid, NSC 401539, NSC 59739
Linear Formula:
O2NC6H4B(OH)2
CAS Number:
Molecular Weight:
166.93

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing