Skip to Content
MilliporeSigma
3-Methyl-5-(2,2,3-trimethyl-3-cyclopenten-1-yl)-4-penten-2-ol analytical standard

3-Methyl-5-(2,2,3-trimethyl-3-cyclopenten-1-yl)-4-penten-2-ol

3-Methyl-5-(2,2,3-trimethyl-3-cyclopenten-1-yl)-4-penten-2-ol analytical standard
Synonym(s):
Ebanol
Empirical Formula (Hill Notation):
C14H24O
CAS Number:
Molecular Weight:
208.34

Products

Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1