Skip to Content
MilliporeSigma
3-Iodo-2-propynyl N-butylcarbamate analytical standard

3-Iodo-2-propynyl N-butylcarbamate

3-Iodo-2-propynyl N-butylcarbamate analytical standard
Linear Formula:
CH3(CH2)3NHCO2CH2C≡Cl
CAS Number:
Molecular Weight:
281.09
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing