Skip to Content
MilliporeSigma
3-Hydroxyphthalic anhydride 98%

3-Hydroxyphthalic anhydride

3-Hydroxyphthalic anhydride 98%
Empirical Formula (Hill Notation):
C8H4O4
CAS Number:
Molecular Weight:
164.11

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing