Skip to Content
MilliporeSigma
3-(Fluorosulfonyl)benzoic acid 95% (HPLC)

3-(Fluorosulfonyl)benzoic acid

3-(Fluorosulfonyl)benzoic acid 95% (HPLC)
Synonym(s):
3-(Fluorosulfonyl)benzoic acid
Empirical Formula (Hill Notation):
C7H5FO4S
CAS Number:
Molecular Weight:
204.18

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing