Skip to Content
MilliporeSigma
3-Chloro-4-nitrotoluene 97%

3-Chloro-4-nitrotoluene

3-Chloro-4-nitrotoluene 97%
Empirical Formula (Hill Notation):
C7H6ClNO2
CAS Number:
Molecular Weight:
171.58
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing