Skip to Content
MilliporeSigma
3-Amino-4-(2-chlorophenyl)-6-nitroquinolin-2(1H)-one British Pharmacopoeia (BP) Reference Standard

3-Amino-4-(2-chlorophenyl)-6-nitroquinolin-2(1H)-one

3-Amino-4-(2-chlorophenyl)-6-nitroquinolin-2(1H)-one British Pharmacopoeia (BP) Reference Standard
Synonym(s):
3-Amino-4-(2-chlorophenyl)-6-nitro-2(1H)-quinolinone
Empirical Formula (Hill Notation):
C15H10ClN3O3
CAS Number:
Molecular Weight:
315.71

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
Special Grade
Showing 1-1 of 1