Skip to Content
MilliporeSigma
3-Acetylpyridine ≥98%, FG

3-Acetylpyridine

3-Acetylpyridine ≥98%, FG
Synonym(s):
Methyl 3-pyridyl ketone
Empirical Formula (Hill Notation):
C7H7NO
CAS Number:
Molecular Weight:
121.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing