Skip to Content
MilliporeSigma
3,4-(Methylenedioxy)aniline 97%

3,4-(Methylenedioxy)aniline

3,4-(Methylenedioxy)aniline 97%
Synonym(s):
5-Amino-1,3-benzodioxole
Empirical Formula (Hill Notation):
C7H7NO2
CAS Number:
Molecular Weight:
137.14
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing