
(3,4-Dimethoxyphenyl)acetonitrile

Synonym(s):
Homoveratronitrile, (3,4-Dimethoxyphenyl)acetonitrile, 3,4-Dimethoxybenzylcyanide
Linear Formula:
(CH3O)2C6H3CH2CN
CAS Number:
Molecular Weight:
177.20
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing