Skip to Content
MilliporeSigma
3,4-Dihydro-2H-pyran 97%

3,4-Dihydro-2H-pyran

3,4-Dihydro-2H-pyran 97%
Synonym(s):
3,4-Dihydro-2H-pyran
Empirical Formula (Hill Notation):
C5H8O
CAS Number:
Molecular Weight:
84.12

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing