Skip to Content
MilliporeSigma
3,4,4′-Trichlorocarbanilide 99%

3,4,4′-Trichlorocarbanilide

3,4,4′-Trichlorocarbanilide 99%
Synonym(s):
1-(4-Chlorophenyl)-3-(3,4-dichlorophenyl)urea, TCC, Triclocarban
Linear Formula:
Cl2C6H3NHCONHC6H4Cl
CAS Number:
Molecular Weight:
315.58

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Agency
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing