Skip to Content
MilliporeSigma
3,3′-Dipropylthiadicarbocyanine iodide suitable for fluorescence, ≥98.0% (TLC)

3,3′-Dipropylthiadicarbocyanine iodide

3,3′-Dipropylthiadicarbocyanine iodide suitable for fluorescence, ≥98.0% (TLC)
Synonym(s):
3-Propyl-2-[5-[3-propyl-2(3H)-benzothiazolylidene]-1,3-pentadienyl]benzothiazolium iodide, DiSC3(5)
Empirical Formula (Hill Notation):
C25H27IN2S2
CAS Number:
Molecular Weight:
546.53

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1