Skip to Content
MilliporeSigma
[3-(3,5-Dichlorophenyl)-2,4-dioxoimidazolidinyl]-N-(methylethyl)carboxamide 97%

[3-(3,5-Dichlorophenyl)-2,4-dioxoimidazolidinyl]-N-(methylethyl)carboxamide

[3-(3,5-Dichlorophenyl)-2,4-dioxoimidazolidinyl]-N-(methylethyl)carboxamide 97%
Synonym(s):
Iprodione
Linear Formula:
Cl2C6H3N2C3H2O2CONHCH(CH3)2
CAS Number:
Molecular Weight:
330.17
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1