Skip to Content
MilliporeSigma
2-Pyridylacetic acid hydrochloride 99%

2-Pyridylacetic acid hydrochloride

2-Pyridylacetic acid hydrochloride 99%
Synonym(s):
2-Pyridineacetic acid hydrochloride
Empirical Formula (Hill Notation):
C7H7NO2 · HCl
CAS Number:
Molecular Weight:
173.60
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing