Skip to Content
MilliporeSigma
2-Propanethiol natural (US), ≥98%, FG

2-Propanethiol

2-Propanethiol natural (US), ≥98%, FG
Synonym(s):
2-Propanethiol
Linear Formula:
(CH3)2CHSH
CAS Number:
Molecular Weight:
76.16

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing