Skip to Content
MilliporeSigma
2-Methyl-1-butanol analytical standard

2-Methyl-1-butanol

2-Methyl-1-butanol analytical standard
Synonym(s):
2-Methyl-1-butanol
Linear Formula:
C2H5CH(CH3)CH2OH
CAS Number:
Molecular Weight:
88.15
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5