Skip to Content
MilliporeSigma
2-Mercaptobenzothiazole certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

2-Mercaptobenzothiazole

2-Mercaptobenzothiazole certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-Benzothiazolethiol, MBT
Empirical Formula (Hill Notation):
C7H5NS2
CAS Number:
Molecular Weight:
167.25

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade