Skip to Content
MilliporeSigma
2-Hydroxypyrimidine hydrochloride 98%

2-Hydroxypyrimidine hydrochloride

2-Hydroxypyrimidine hydrochloride 98%
Synonym(s):
2(1H)-Pyrimidinone hydrochloride, 2-Pyrimidinol hydrochloride
Empirical Formula (Hill Notation):
C4H4N2O · HCl
CAS Number:
Molecular Weight:
132.55
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing